Learn More
Allyl acetate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 041353AD
Description
Ally acetate is used in industrial intermediate. Used in synthesis of colophony and bond.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Allyl acetate | |
6°C | |
103°C to 104°C | |
1.4042 | |
50 mL | |
14,285 | |
Soluble in ethanol. Miscible with ethanol and ether. Slightly soluble in water. | |
CC(=O)OCC=C | |
100.117 | |
100.12 | |
Liquid |
591-87-7 | |
0.928 g/mL | |
C5H8O2 | |
MFCD00008721 | |
UN2333 | |
allyl acetate, 3-acetoxypropene, acetic acid, 2-propenyl ester, 2-propenyl acetate, 2-propenyl ethanoate, 3-acetoxy-1-propene, allylacetate, acetic acid, allyl ester, 2-propenyl methanoate, unii-e4u5e5990i | |
FWZUNOYOVVKUNF-UHFFFAOYSA-N | |
prop-2-enyl acetate | |
11584 | |
99% |
Chemical Identifiers
591-87-7 | |
100.117 | |
FWZUNOYOVVKUNF-UHFFFAOYSA-N | |
11584 | |
CC(=O)OCC=C |
C5H8O2 | |
MFCD00008721 | |
allyl acetate, 3-acetoxypropene, acetic acid, 2-propenyl ester, 2-propenyl acetate, 2-propenyl ethanoate, 3-acetoxy-1-propene, allylacetate, acetic acid, allyl ester, 2-propenyl methanoate, unii-e4u5e5990i | |
prop-2-enyl acetate |
Safety and Handling
GHS H Statement
H225-H301-H312
Highly flammable liquid and vapor.
Toxic if swallowed.
Harmful in contact with skin.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H331-H312-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALLYL ACETATE
EINECSNumber : 209-734-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only