Alluviam HazMasterG3™ CBRNE/IED Threat Identification System and HazMapper™ Threat-Mapping Software
Choose between comprehensive mobile HazMat/CBRNE/IED decision aid and threat-mapping software, or integrate the two
$401.31 - $3,493.65
Specifications
Certifications/Compliance U.S.DHS, DoD COE/JTA
View More Specs
Includes: First year of annual maintenance and support.
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes For Use With (Application) Product Type Price Quantity  
Catalog Number Mfr. No. Includes For Use With (Application) Product Type Price Quantity  
19-130-6002 Alluviam Llc
HMG3DPPC
Software for handheld computing devices comes on SD/MMC memory card with CD-ROM backup, CD-ROM contains program files, user manual and includes 37+ responder reference guides, first year of annual maintenance and support Identify an extremely broad range of unknown chemical agents, radiological isotopes, complex improvised explosive device (IED) threats, detonators, intermodal containers and road and rail hazards Handheld pocket PC device Each for $3,493.65
19-130-6013 Alluviam Llc
HMG3AS
First year of annual maintenance and support For HazMasterG3 CBRNE identification system Additional year of software/data maintenance support Each for $401.31
19-130-6005 Alluviam Llc
HMG3WIN
First year of annual maintenance and support, with software installer on CD-ROM with program files, user manual and 37+ supplemental first-responder reference guides For Windows™ HazMasterG3 software Each for $2,820.22
Specifications
Description
HazMasterG3 CBRNE/IED Threat Identification System and Decision Support
Can identify an extremely broad range of unknown chemical agents, radiological isotopes, complex improvised explosive device (IED) threats, detonators, intermodal containers, and road and rail hazards. Provides an industry-leading volume of critical response information. U. S. DHS certified as an approved Anti-Terror Technology.
  • Can quickly identify the specific agents most likely to cause observed symptoms and provide immediate access to all relevant response information
  • Lets you mix an arbitrary number of agents, and predicts the most probable precursor outcomes
  • Delivers critical response guidance for 117,000+ agents (TICs, TIMs, CW agents), radiological isotopes, bio-weapons, bio-terror agents, explosives, and IEDs
  • Tactical TIER 2 facility preplan support for 60,000+ facilities nationwide
  • Provides comprehensive detonator identification for 7000+ varieties of commerical/military detonators, X-ray and disruptor working aid calculators, and IED improvised trigger component identifiation for nearly 10,000 components
  • Offers reach back access to 2,000,000+ materials and trade names MSDS repository
  • Contains over 32 data sources integrated together
  • Supports 8 different platforms, from smart cell phones, to hand held and laptop/desktop computers
HazMapper Threat-Mapping Software
Delivers street-level plume and tactical threat overlays for HAZMAT/CBRNE/IED incidents in a fast and easy GIS situational awareness tool. Automatically displays HazMasterG3 isolation and evacuation distances for incidents involving chemical, radiological, improvised explosive devices (IEDs) and improvised nuclear devices (INDs) along with ALOHA or HPAC plumes.
  • Requires no specialized GIS training
  • Provides comprehensive, geographical situational-awareness as HAZMAT/CBRNE/IED incidents unfold
  • Can provide a common operating picture to remote operation centers and others involved in incident response
  • Works with a wide range of GPS receivers, and compatible with a wide range of industry standard GIS system overlays
  • Hands free moving map capability keeps users centered on the map as they move
  • Provides seamless integration with HazMasterG3
Order Info
NIMS and ICS compatible. Runs in any environment that supports HPAC 4.0. Supported platforms include Microsoft Windows™ (95-XP), Pocket PC, Palm OS™ (3-5.x), Macintosh™ OS-X, Linux, Solaris, HP/UX, and smart cell phones. (For configurations not described below, contact your Fisher Scientific™ Safety Customer Service Representative.) Windows-compatible software ships with software installer on a CD-ROM which contains program files, user manual, and 37+ supplemental first-responder reference guides. Software for handheld computing devices comes on SD/MMC memory card with CD-ROM backup. System requirements: Approx. 10 MB of disk space.
Optional Mil-Spec case is water/dust/shock/crush proof.
Certifications
Has earned U. S. DHS certification and designation as an approved Anti-Terror Technology.
Compliance
DoD COE/JTA compliant. Special ruggedized recon device enjoys IP67 protection; exceeds military specifications for drop, vibration, and both high and low temperature operation.
Description & Specifications
Specifications
U.S.DHS, DoD COE/JTA