Please login to your online account to display your discounted pricing

Alloxan monohydrate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  215370250

 View more versions of this product

Catalog No. AC215370250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 160.09
Physical Form Crystalline Powder
Quantity 25g
Color Off-white to beige-yellow
RTECS Number UW0492000
Beilstein 24, 500
Merck Index 10, 276
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
CAS Min % 97.5
CAS 2244-11-3
CAS 50-71-5
Chemical Name or Material Alloxan monohydrate
Molecular Formula C4H2N2O4·H2O
MDL Number MFCD00149399
Assay Percent Range 98%
Packaging Glass bottle
Synonym 2,4,5,6(1H,3H)-Pyrimidinetetrone monohydrate
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Alloxan monohydrate
Name Note 98%
CAS 2244-11-3
CAS Min % 97.5
CAS Max % 100.0
CAS 50-71-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H2N2O4·H2O
Formula Weight 160.09
MDL Number MFCD00149399
Synonym 2,4,5,6(1H,3H)-Pyrimidinetetrone monohydrate
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

SDS & Safety Documents