Please login to your online account to display your discounted pricing

Alloxan monohydrate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Alloxan monohydrate
Name Note 98%
CAS 2244-11-3
CAS Min % 97.5
CAS Max % 100.0
CAS 50-71-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H2N2O4·H2O
Formula Weight 160.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC215370250

  SDS
thermo fisher scientific
215370250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 160.09
Quantity 25g
Color Off-white to beige-yellow
RTECS Number UW0492000
Beilstein 24, 500
Merck Index 10, 276
CAS Max % 100.0
Name Note 98%
CAS 2244-11-3
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Alloxan monohydrate
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C4H2N2O4·H2O
MDL Number MFCD00149399
Synonym 2,4,5,6(1H,3H)-Pyrimidinetetrone monohydrate
CAS 50-71-5
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.