Please login to your online account to display your discounted pricing

Allophycocyanin Mouse, Clone: eBioAPC-6A2, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $404.55

Specifications

Antigen Allophycocyanin
Clone eBioAPC-6A2
Host Species Mouse
Gene Alias Anti-APC
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013032

 
life technologies
LS14417081
50μg Each for $114.55

5013033

 
life technologies
LS14417085
500μg Each for $404.55
Description & Specifications

Specifications

Antigen Allophycocyanin
Clone eBioAPC-6A2
Host Species Mouse
Gene Alias Anti-APC
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBioAPC-6A2 monoclonal antibody reacts with allophycocyanin (APC) and APC-Cy7, with no crossreactivity to fluorescein (FITC) or phycoerythrin (PE). When using eBioAPC-6A2 as a second-step antibody, in combination with an appropriate third step, to bind a APC-conjugated primary antibody, there is no decrease in the fluorescent intensity of the directly conjugated APC antibody. Therefore no interference or quenching of the APC fluorochrome is observed. The eBioAPC-6A2 can be used for separation of cells labeled with APC-conjugated antibodies or for staining.

Please note that the optimal concentration for this antibody will vary depending on the density of the primary APC-conjugated antibody and the application tested.