Please login to your online account to display your discounted pricing

Allophycocyanin Mouse, Biotin, Clone: eBioAPC-6A2, eBioscience™

Mouse Monoclonal Antibody

$71.05 - $158.05

Specifications

Antigen Allophycocyanin
Clone eBioAPC-6A2
Host Species Mouse
Gene Alias Anti-APC
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122542

 
life technologies
LS13417082
100μg Each for $158.05

5011849

 
life technologies
LS13417080
25μg Each for $71.05
Description & Specifications

Specifications

Antigen Allophycocyanin
Clone eBioAPC-6A2
Host Species Mouse
Gene Alias Anti-APC
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The eBioAPC-6A2 monoclonal antibody reacts with allophycocyanin (APC) and APC-Cy7, with no crossreactivity to fluorescein (FITC) or phycoerythrin (PE). When using eBioAPC-6A2 as a second-step antibody, in combination with an appropriate third step, to bind a APC-conjugated primary antibody, there is no decrease in the fluorescent intensity of the directly conjugated APC antibody. Therefore no interference or quenching of the APC fluorochrome is observed. The eBioAPC-6A2 can be used for separation of cells labeled with APC-conjugated antibodies or for staining.

Please note that the optimal concentration for this antibody will vary depending on the density of the primary APC-conjugated antibody and the application tested.