missing translation for 'onlineSavingsMsg'
Learn More
missing translation for 'pleaseLogIn'

Alfa Aesar™ Alloferon 2

For cellular and molecular biology applications

Specifications

Product Type Alloferon 2
Molecular Weight 1128.17
Storage Requirements -30° to -10°C
Format Solid
Sequence Gly-Val-Ser-Gly-His-Gly-Gln-His-Gly-Val-His-Gly
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAJ66965EXD Alfa Aesar™
J66965EXD
0.5mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66965MCR Alfa Aesar™
J66965MCR
1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Specifications

Alloferon 2
-30° to -10°C
Gly-Val-Ser-Gly-His-Gly-Gln-His-Gly-Val-His-Gly
HPLC
1128.17
Solid
RUO