Please login to your online account to display your discounted pricing

Alfa Aesar™ Alloferon 1

For cellular and molecular biology applications

Specifications

Product Type Alloferon 1
Molecular Weight 1265.31
Storage Requirements -30° to -10°C
Format Solid
Sequence His-Gly-Val-Ser-Gly-His-Gly-Gln-His-Gly-Val-His-Gly
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66879EXD

 
alfa aesar™
J66879EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66879MCR

 
alfa aesar™
J66879MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Alloferon 1
Molecular Weight 1265.31
Storage Requirements -30° to -10°C
Format Solid
Sequence His-Gly-Val-Ser-Gly-His-Gly-Gln-His-Gly-Val-His-Gly
Regulatory Status RUO
Purification Method HPLC