Please login to your online account to display your discounted pricing

Alfa Aesar™ Alloferon 1


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66879MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66879MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Alloferon 1
Regulatory Status RUO
Format Solid
Storage Requirements -30° to -10°C
Quantity 1mg
Molecular Weight 1265.31
Sequence His-Gly-Val-Ser-Gly-His-Gly-Gln-His-Gly-Val-His-Gly
Purification Method HPLC