missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Allegro™ Bitrex Fit Test Kit: Solution

For use with Allegro Bitrex Fit Test Kit (2041).

Manufacturer:  Allegro™ 204111K

 View more versions of this product

Catalog No. 19-088-509



Description

Description

Bitrex Sensitivity Solution and Bitrex Test Solution

  • Box of 6 glass ampules each
  • With pre-drilled plastic cap for easy pouring
  • Plastic protection from broken glass
  • One-time use
  • Averages 3 to 5 fit tests per tube
  • Contains 2.5cc of solution

 

Specifications

Specifications

OSHA
Sensitivity Solution
Sensitivity solution
Allegro Bitrex Fit Test Kit (2041)
6/Pk.
SDS
Documents
Product Certifications