Allegro™ Banana Oil Fit-Test Kits
Ensure a proper respirator fit
Manufacturer: Allegro™ 0203
Description
- Banana oil (isoamyl acetate) is the substance originally used for testing respirator fit; its strong, recognizable odor quickly indicates the lack of proper fit
- Available in two fit test kits and Fit Check Ampules that provide a fast and easy method of on-the-spot qualitative fit testing
Order Info
The Standard Kit comes in a handy carry bag and includes two 1 oz. (30mL) bottles of testing agent, a vinyl test enclosure, measuring pipette, and paper towels. The Deluxe Kit comes in a convenient storage case and contains everything necessary to perform a proper fit test, inlcuding an 8 1oz. (30mL) bottles of testing solution, a fit test tent, jars, paper towels, disposable bags, and other necessary components. Replacement banana oil solution is available in 1 oz. (30mL) bottles.
Specifications
Used for testing respirator fit | |
Two 1 oz. bottles of test agents, vinyl test enclosure, measuring pipette and paper towels in a handy carry bag | |
Standard Banana Oil Fit-Test Kit | |
1 Ea. |