Please login to your online account to display your discounted pricing

Alfa Aesar™ Allatostatin IV

For cellular and molecular biology applications

Specifications

Product Type Allatostatin IV
Molecular Weight 969.09
Storage Requirements -30° to -10°C
Sequence Asp-Arg-Leu-Tyr-Ser-Phe-Gly-Leu-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66648MA

 
alfa aesar™
J66648MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66648MB

 
alfa aesar™
J66648MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Allatostatin IV
Molecular Weight 969.09
Storage Requirements -30° to -10°C
Sequence Asp-Arg-Leu-Tyr-Ser-Phe-Gly-Leu-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC