Please login to your online account to display your discounted pricing

Alfa Aesar™ Allatostatin IV


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66648MB

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66648MB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 25mg
Storage Requirements -30° to -10°C
Product Type Allatostatin IV
Sequence Asp-Arg-Leu-Tyr-Ser-Phe-Gly-Leu-NH2
Molecular Weight 969.09
Purification Method HPLC
Regulatory Status RUO
Format Solid