Please login to your online account to display your discounted pricing

Alfa Aesar™ Allatostatin III

For cellular and molecular biology applications

Specifications

Product Type Allatostatin III
Molecular Weight 899.00
Storage Requirements -30° to -10°C
Sequence Gly-Gly-Ser-Leu-Tyr-Ser-Phe-Gly-Leu-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66599LB0

 
alfa aesar™
J66599LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66599MCR

 
alfa aesar™
J66599MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Allatostatin III
Molecular Weight 899.00
Storage Requirements -30° to -10°C
Sequence Gly-Gly-Ser-Leu-Tyr-Ser-Phe-Gly-Leu-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC