Please login to your online account to display your discounted pricing

Alfa Aesar™ Allatostatin II

For cellular and molecular biology applications

Specifications

Product Type Allatostatin II
Molecular Weight 1067.19
Storage Requirements -30° to -10°C
Format Solid
Sequence Gly-Asp-Gly-Arg-Leu-Tyr-Ala-Phe-Gly-Leu-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66779LB0

 
alfa aesar™
J66779LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66779MCR

 
alfa aesar™
J66779MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Allatostatin II
Molecular Weight 1067.19
Storage Requirements -30° to -10°C
Format Solid
Sequence Gly-Asp-Gly-Arg-Leu-Tyr-Ala-Phe-Gly-Leu-NH2
Regulatory Status RUO
Purification Method HPLC