Please login to your online account to display your discounted pricing

Alfa Aesar™ Allatostatin I

For cellular and molecular biology applications

Specifications

Product Type Allatostatin I
Molecular Weight 1335.51
Storage Requirements -30° to -10°C
Sequence Ala-Pro-Ser-Gly-Ala-Gln-Arg-Leu-Tyr-Gly-Phe-Gly-Leu-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66635LB0

 
alfa aesar™
J66635LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66635MA

 
alfa aesar™
J66635MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Allatostatin I
Molecular Weight 1335.51
Storage Requirements -30° to -10°C
Sequence Ala-Pro-Ser-Gly-Ala-Gln-Arg-Leu-Tyr-Gly-Phe-Gly-Leu-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC