missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Allantoin, USP, 98.5-101%, Spectrum™ Chemical
A2721, 97-59-6, C4H6N4O3
Supplier: Spectrum Chemical Mfg Cor A272125KGBL
Description
Spectrum™ Chemical Allantoin, USP promotes cell proliferation and wound healing and has a soothing, anti-irritant, and skin protectant effect. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
97-59-6 | |
0.1% | |
Fiber Drum | |
25 kg | |
NC(=O)NC1NC(=O)NC1=O | |
158.12 | |
USP |
100% | |
0.1% | |
C4H6N4O3 | |
POJWUDADGALRAB-UHFFFAOYNA-N | |
(2,5-dioxoimidazolidin-4-yl)urea | |
98.5 to 101% |
Chemical Identifiers
97-59-6 | |
158.12 | |
(2,5-dioxoimidazolidin-4-yl)urea |
C4H6N4O3 | |
POJWUDADGALRAB-UHFFFAOYNA-N | |
NC(=O)NC1NC(=O)NC1=O |