missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

all-trans-Retinol, 95%, ACROS Organics™

$72.95 - $352.84

Chemical Identifiers

CAS 68-26-8
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C20H30O
Formula Weight 286.46
MDL Number MFCD00001552
Synonym Vitamin A
Chemical Name or Material all-trans-Retinol, 95%
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC347070010 SDS Thermo Fisher Scientific
AC347070010
Glass bottle 1g Each for $352.84
AC347072500 SDS Thermo Fisher Scientific
AC347072500
Glass bottle 250mg Each for $142.46
AC347075000 SDS Thermo Fisher Scientific
AC347075000
Glass bottle 500mg Each for $223.33
AC347071000 SDS Fisher Scientific Acros
347071000
Glass bottle 100mg Each for $72.95
Specifications Description & Specifications

Chemical Identifiers

68-26-8
100.0
C20H30O
MFCD00001552
all-trans-Retinol, 95%
94.0
95%
286.46
Vitamin A
≥94%

Specifications

68-26-8
100.0
Glass bottle
95%
286.46
06, III, 2787
Vitamin A
1g
≥94%
94.0
Authentic
Powder and Lumps
C20H30O
MFCD00001552
15, 10211
Yellow to orange
all-trans-Retinol, 95%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Suspected of damaging the unborn child.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 200-683-7

RTECSNumber : VH6750000

TSCA : TSCA