missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

all cis-8,11,14-Eicosatrienoic Acid, TCI America™

Manufacturer:  TCI America E064010MG

 View more versions of this product

Catalog No. E064010MG



Specifications

Specifications

all cis-8,11,14-Eicosatrienoic Acid
Yellow
1783-84-2
MFCD00065721
HOBAELRKJCKHQD-QNEBEIHSSA-N
(8Z,11Z,14Z)-icosa-8,11,14-trienoic acid
5280581
306.49
≥98.0% (GC)
200°C
10mg
C20H34O2
dihomo-gamma-linolenic acid, dgla, gamma-homolinolenic acid, 8,11,14-eicosatrienoic acid, homo-gamma-linolenic acid, cis-8,11,14-eicosatrienoic acid, bishomo-gamma-linolenic acid, z,z,z-8,11,14-eicosatrienoic acid, all-cis-8,11,14-eicosatrienoic acid, 8z,11z,14z-icosa-8,11,14-trienoic acid
CCCCCC=CCC=CCC=CCCCCCCC(=O)O
306.49
CHEBI:53486
Clear Liquid at 20°C

Chemical Identifiers

1783-84-2
306.49
HOBAELRKJCKHQD-QNEBEIHSSA-N
5280581
(8Z,11Z,14Z)-icosa-8,11,14-trienoic acid
C20H34O2
MFCD00065721
dihomo-gamma-linolenic acid, dgla, gamma-homolinolenic acid, 8,11,14-eicosatrienoic acid, homo-gamma-linolenic acid, cis-8,11,14-eicosatrienoic acid, bishomo-gamma-linolenic acid, z,z,z-8,11,14-eicosatrienoic acid, all-cis-8,11,14-eicosatrienoic acid, 8z,11z,14z-icosa-8,11,14-trienoic acid
CHEBI:53486
CCCCCC=CCC=CCC=CCCCCCCC(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0609

RTECSNumber : JX3880000

TSCA : No

Recommended Storage : Freezer (-20 °C)

Documents