missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkylphenol Target Analyte Mix Solution for DIN EN ISO 18857-2 in Acetone, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| CAS | 67-64-1 |
| Flash Point | -17.0°C - Closed Cup - Solvent |
| Type | Analytical Standard |
| Quantity | 10 mL |
| Grade | Analytical Standard |