missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alkaline-Iodide Reagent, Winkler Formulation, for Dissolved Oxygen (DO) Analysis, Ricca Chemical

For Analysis of Dissolved Oxygen (DO)

$136.85 - $414.39

Chemical Identifiers

CAS 1310-73-2,7732-18-5,7681-11-0
CAS Min % 46.0,45.51,6.83
CAS Max % 48.0,47.37,7.11
Physical Form Liquid
Chemical Name or Material Alkaline-Iodide Reagent, Winkler Formulation
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
530-1 Ricca Chemical Company
RI5301
Poly natural 4L Each for $414.39
530-32 Ricca Chemical Company
RI53032
Poly natural 1L Each for $136.85
R0530000100 Ricca Chemical Company
RIR0530000100A
Poly natural 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1310-73-2,7732-18-5,7681-11-0
48.0,47.37,7.11
Alkaline-Iodide Reagent, Winkler Formulation
46.0,45.51,6.83
Liquid

Specifications

1310-73-2,7732-18-5,7681-11-0
48.0,47.37,7.11
Liquid
4L
46.0,45.51,6.83
Poly natural
Clear colorless
Alkaline-Iodide Reagent, Winkler Formulation