Please login to your online account to display your discounted pricing

Alkaline Iodide Azide Solution II, For Dissolved Oxygen (Concentrations Greater than 15mg/L), LabChem™

$670.26 - $670.26

Chemical Identifiers

Chemical Name or Material Alkaline Iodide Azide Solution II
CAS 7681-82-5
CAS Max % 48.23
CAS 7732-18-5
CAS Max % 29.47
CAS 1310-73-2
CAS Max % 21.76
CAS 26628-22-8
CAS Max % 0.54
Name Note For Dissolved Oxygen (Concentrations Greater than 15mg/L)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC106902

 
lab chem
LC106902
Amber Poly 1L Each for $670.26
Description & Specifications

Specifications

Formula Weight 65.01
Solubility Information Soluble in water
Name Note For Dissolved Oxygen (Concentrations Greater than 15mg/L)
Identification Passes Test
CAS 7732-18-5
CAS Max % 48.23
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Physical Form Liquid
Health Hazard 2 GHS H Statement
Harmful if swallowed or in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Color Clear, colorless
Packaging Amber Poly
CAS 1310-73-2
CAS 7681-82-5
Linear Formula NaN3
Health Hazard 1 Danger
Chemical Name or Material Alkaline Iodide Azide Solution II
CAS Max % 21.76
CAS 26628-22-8
CAS Max % 29.47
CAS Max % 0.54
UN Number UN1824
Quantity 1L
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed or in contact with skin.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

Room Temperature