missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alkaline-Iodide-Azide, for Dissolved Oxygen (DO) Analysis, Ricca Chemical

For Analysis of Dissolved Oxygen (DO)

Chemical Identifiers

CAS 7681-82-5,7732-18-5,1310-73-2,26628-22-8
CAS Min % 42.0,28.13,27.05,0.56
CAS Max % 44.0,29.28,28.15,0.59
Physical Form Liquid
Chemical Name or Material Alkaline-Iodide-Azide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
541-16 Ricca Chemical Company
RI54116
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
541-32 Ricca Chemical Company
RI54132
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7681-82-5,7732-18-5,1310-73-2,26628-22-8
44.0,29.28,28.15,0.59
Alkaline-Iodide-Azide
42.0,28.13,27.05,0.56
Liquid

Specifications

Liquid
500mL
44.0,29.28,0.59,28.15
Poly natural
Clear to turbid
7681-82-5,7732-18-5,1310-73-2,26628-22-8
27.05,42.0,28.13,0.56
Alkaline-Iodide-Azide