missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Alkali Metals, plasma standard solution, Specpure™, Ba,Be,Ca,Cs,K,Li,Mg,Na,Rb,Sr at 100μg/mL

Manufacturer:  Alfa Aesar™ 44428AE

 View more versions of this product

Catalog No. AA44428AE



Specifications

Specifications

Alkali Metals, plasma standard solution
MFCD08061357
Liquid
100mL
Ba,Be,Ca,Cs,K,Li,Mg,Na,Rb,Sr at 100μg/mL
UN3264
Specpure™
Matrix: 5% HNO3
Documents
Promotions

Promotions