missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Alkali Blue 4B

$48.62 - $170.54

Chemical Identifiers

CAS 62152-67-4
Molecular Formula C32H28N3NaO4S
Formula Weight 573.65
MDL Number MFCD00078891
Synonym C.I. 42750
Chemical Name or Material Alkali Blue 4B
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1796614 SDS Alfa Aesar™
A1796614
25g Each for $48.62
AAA1796622 SDS Alfa Aesar™
A1796622
100g Each for $170.54
Specifications Description & Specifications

Chemical Identifiers

62152-67-4
573.65
C.I. 42750
C32H28N3NaO4S
MFCD00078891
Alkali Blue 4B

Specifications

62152-67-4
573.65
C.I. 42750
Alkali Blue 4B
C32H28N3NaO4S
MFCD00078891
25g

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning