Invitrogen™ ALK Recombinant Human Protein DFS Item
Anaplastic Lymphoma Receptor Tyrosine Kinase (ALK) belongs to the insulin receptor superfamily.
Manufacturer:  Invitrogen™ LSPV4185
 View more versions of this product
Catalog No. PV4185
Description
Description
ALK is vital for brain development. Mutations, rearrangements, and amplifications in the ALK gene have been found in tumors, including anaplastic large cell lymphoma, neurblastoma, and non-small cell lung cancer. Chromosomal rearrangement is the most common genetic alteration.
Specifications
Specifications
NP_004295
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
89.9kDa
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
CD246, NBLST3
Baculovirus
Human
GST Tag
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
238
1mg
Active
Kinase Assay
ALK Kinase Family
Yes
Documents