ALK [C1156Y] Recombinant Human Protein, Invitrogen™ DFS Item
Human ALK C1156Y, internal fragment, amino acids L1066-S1437 (as in NCBI/Protein entry NP_004295.2), C1156Y mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
$614.00 - $9,380.00
Specifications
Host Species Human
Protein Tag GST Tag, His Tag
Recombinant Yes
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV6157 DFS Item
View Documents
Life Technologies
LSPV6157
10μg
Each for $614.00
PV6158 DFS Item
View Documents
Life Technologies
LSPV6158
100μg
Each for $2,095.00
PV6159 DFS Item
View Documents
Life Technologies
LSPV6159
1mg
Each for $9,380.00
Description
Description
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Human
GST Tag, His Tag
Yes
SDS