Please login to your online account to display your discounted pricing

Alizarin Red S, Reagent Grade, LabChem™

$106.20 - $106.20

Chemical Identifiers

Chemical Name or Material Alizarin Red S
CAS 130-22-3
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C14H7NaO7S
Formula Weight 342.27
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC105908

 
lab chem
LC105908
25g Each for $106.20
Description & Specifications

Specifications

Formula Weight 342.27
Chemical Name or Material Alizarin Red S
Quantity 25g
Purity Grade Reagent Grade
Physical Form Powder
Linear Formula C14H7NaO7S
CAS 130-22-3
Identification Passes Test
Health Hazard 1 Warning
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Store in a well-ventilated place.
Keep container tightly closed.
If on skin: Wash skin with plenty of soap and water.
If skin irritation occurs: Get medical advice.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Sodium oxide Carbon monoxide Carbon dioxide Sulfur compounds
Color Reddish-orange
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Store in a well-ventilated place.
Keep container tightly closed.
If on skin: Wash skin with plenty of soap and water.
If skin irritation occurs: Get medical advice.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation and serious eye irritation.
May cause respiratory irritation.

Room Temperature