Please login to your online account to display your discounted pricing

Alfa Aesar™ Alizarin Complexone, may cont. up to 10% water

$101.57 - $343.54

Chemical Identifiers

Chemical Name or Material Alizarin Complexone
CAS 3952-78-1
Molecular Formula C19H15NO8
Formula Weight 385.33
MDL Number MFCD00001202
Beilstein 2190028
Synonym Alizarin Fluorine Blue; Alizarin-3-methyliminodiacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1669903

 
alfa aesar™
A1669903
1g Each for $101.57

AAA1669906

 
alfa aesar™
A1669906
5g Each for $343.54
Description & Specifications

Specifications

Formula Weight 385.33
Chemical Name or Material Alizarin Complexone
Quantity 1g
TSCA TSCA
Assay Percent Notes may cont. up to 10% water
Molecular Formula C19H15NO8
MDL Number MFCD00001202
CAS 3952-78-1
Beilstein 2190028
Synonym Alizarin Fluorine Blue; Alizarin-3-methyliminodiacetic acid
Melting Point 180° to 182°C
RTECS Number AH0585000

Applications
Alizarin is the main ingredient for the manufacture of the madder lake pigments known to painters as Rose madder and Alizarin crimson. A notable use of alizarin in modern times is as a staining agent in biological research because it stains free calcium and certain calcium compounds a red or light purple color. Alizarin Red is used in a biochemical assay to determine, quantitatively by colorimetry, the presence of calcific deposition by cells of an osteogenic lineage.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.