Please login to your online account to display your discounted pricing

Aliquat™ 336, average MW 442, ACROS Organics™

$90.73 - $351.78

Chemical Identifiers

Chemical Name or Material Aliquat™ 336
Name Note Average M.W. 442
CAS 63393-96-4
Assay 88.2 to 90.6%, 112.0 to 115.0 (quaternary value)
Formula Weight 442
MDL Number MFCD00011862
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC194970025

  SDS
thermo fisher scientific
AC194970025
Glass bottle 2.5L Each for $351.78

AC194970010

  SDS
fisher scientific acros
194970010
Glass bottle 1L Each for $252.98

AC194972500

  SDS
thermo fisher scientific
194972500
Glass bottle 250mL Each for $90.73
Description & Specifications

Specifications

Physical Form Viscous Liquid
Formula Weight 442
Quantity 2.5L
Color Amber
Assay 88.2 to 90.6%, 112.0 to 115.0 (quaternary value)
Density 0.8800g/mL
Fieser 05,460; 06,404; 07,380; 13,192
Name Note Average M.W. 442
Packaging Glass bottle
Refractive Index 1.4655 to 1.4715 (20°C, 589nm)
Health Hazard 2 GHS P Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material Aliquat™ 336
MDL Number MFCD00011862
CAS 63393-96-4
Water 5% max.
Health Hazard 1 Danger

Phase transfer catalyst

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.