Thermo Scientific™ aLICator Ligation Independent Cloning and Expression System
Get fast and efficient ligation-independent cloning and tight regulation of gene expression in E. coli. with this cloning and expression system.
$215.00 - $272.00
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
FERK1291 Thermo Scientific™
K1291
aLICator LIC Cloning and Expression Set 2 (All-in-One/WQ) Each for $272.00
FERK1241 Thermo Scientific™
K1241
Each for $218.00
FERK1251 Thermo Scientific™
K1251
Each for $215.00
FERK1261 Thermo Scientific™
K1261
Each for $218.00
FERK1271 Thermo Scientific™
K1271
Each for $272.00
Specifications
Description
aLICator LIC Cloning and Expression System is designed for fast and efficient ligation independent cloning and tight regulation of gene expression in E. coli. The pLATE bacterial expression vectors are designed for high levels of target protein expression in concert with minimal background (uninduced) expression, which permits expression of proteins that are toxic to E. coli cells. To streamline and facilitate the process of insert cloning into the expression vector, the aLICator system uses directional LIC cloning technology, a rapid procedure that provides high-cloning efficiencies.
The tightly regulated expression and fast, efficient directional cloning makes the aLICator LIC Cloning and Expression System the best choice for routine and toxic gene cloning and expression in E. coli.
Highlights
  • High efficiency LIC cloning
  • Tight control of gene expression
  • High yield expression
  • Versatile -tagged or untaged protein expression with tag removal option