Please login to your online account to display your discounted pricing
Alfa Aesar™ Yttria Stabilized Zirconium Oxide Cylindrical Crucibles
$224.73 - $1,008.23
Specifications
Product Type Yttria Stabilized Zirconium Oxide Cylindrical Crucibles
Quantity 1 Piece
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Height (Metric) Volume (English) Price Quantity    
AA41273KF
alfa aesar
41273KF
20mm 37mm 5mL Each for $224.73
AA41274KF
alfa aesar
41274KF
24mm 43mm 10mL Each for $326.32
AA41275KF
alfa aesar
41275KF
37mm 72mm 50mL Each for $421.76
AA41276KF
alfa aesar
41276KF
45mm 90mm 100mL Each for $518.74
AA41277KF
alfa aesar
41277KF
55mm 112mm 200mL Each for $1,008.23
Description & Specifications
Specifications
Product Type Yttria Stabilized Zirconium Oxide Cylindrical Crucibles
Quantity 1 Piece