Please login to your online account to display your discounted pricing
Alfa Aesar™ Wig-L-Bug™ Mixing/Grinding Mills
Easy to prepare and mix samples for IR analysis
$1,563.12 - $1,563.12
Specifications
Product Type Mixing/Grinding Mill
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    
AA42627KT
alfa aesar
42627KT
Cover Each for $1,563.12
AA44204KT
alfa aesar
44204KT
Not Applicable CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA44604KT
alfa aesar
44604KT
Without cover CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Product Type Mixing/Grinding Mill
  • Easy-to-use icon-driven control panel
  • Membrane keypad is easily cleaned and disinfected
  • Safety cover shields capsules, reduces noise (no cover for 44604)
  • Speeds of 3,800, 4,200, 4,600, and 4,800rpm
  • Accurate digital circuitry with LED readout of time and speed
  • Vials and pestles sold separately
Model Specific Features:
Heavy Duty version (42627)
  • Useful for mixing and grinding that require constant and long periods of operation
  • Has 30 minute timer and fan-cooled motor
  • Comes with vial adapter for 1.27cm (0.5 in.) diameter × 2.54cm (1 in.) long vials