Please login to your online account to display your discounted pricing
Alfa Aesar™ USA Standard Sieves: 200 to 270 Mesh
Brass frame and Stainless-steel mesh sieves
$143.01 - $252.08
Specifications
Product Type Sieve
Material Stainless steel
Certifications/Compliance ASTM E11
View More Specs
Products
Catalog Number Mfr. No. Depth (Metric) Description Diameter (Metric) Outer Price Quantity    
AA39991NK
alfa aesar
39991NK
7.62cm 200 Mesh 30.4cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39991NR
alfa aesar
39991NR
5.08cm 200 Mesh 20.3cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39991ON
alfa aesar
39991ON
2.54cm 200 Mesh 7.62cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39992NK
alfa aesar
39992NK
7.62cm 230 Mesh 30.4cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39992NR
alfa aesar
39992NR
5.08cm 230 Mesh 20.3cm Each for $252.08
AA39992ON
alfa aesar
39992ON
2.54cm 230 Mesh 7.62cm Each for $143.01
AA39993NK
alfa aesar
39993NK
7.62cm 270 Mesh 30.4cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39993NR
alfa aesar
39993NR
5.08cm 270 Mesh 20.3cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA39993ON
alfa aesar
39993ON
2.54cm 270 Mesh 7.62cm Each for $154.11
Description & Specifications
Specifications
Product Type Sieve
Material Stainless steel
Certifications/Compliance ASTM E11
  • Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
  • Sieves are full height
Sieve Depths
  • 3 in. diameter sieves are 1 in. deep
  • 8 in. diameter sieves are 2 in. deep
  • 12 in.diameter sieves are 3 in. deep