Thermo Scientific™ USA Standard Sieves, 18 Mesh
Brass frame and Stainless-steel mesh sieves
Supplier: Thermo Scientific™ 43658NK
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
- Sieves are full height
- 3 in. diameter sieves are 1 in. deep
- 8 in. diameter sieves are 2 in. deep
- 12 in.diameter sieves are 3 in. deep
Specifications
18 Mesh | |
3 in. | |
Sieve | |
12 in. |
30.4cm | |
7.62cm | |
ASTM E11 | |
Stainless Steel |