Please login to your online account to display your discounted pricing
Alfa Aesar™ Thin Film Spin Coater Accessory, Chucks
For use with Thin Film Spin Coater
Specifications
Product Type Chuck
For Use With (Equipment) Thin Film Spin Coater
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Price Quantity    
AA43924KT
alfa aesar
43924KT
2.54cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Product Type Chuck
For Use With (Equipment) Thin Film Spin Coater
  • Machined to close tolerances to assure smooth spinning process and uniform coating
  • Chuck selection should be based on substrate size and geometry
  • Vacuum chuck used to hold substrate for coating