missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel plating solution, electroless

For Plating On Silicon, Germanium, Gallium Arsenide

$96.49 - $96.49

Chemical Identifiers

Physical Form Liquid
MDL Number MFCD00011137
Chemical Name or Material Nickel plating solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA44069AK SDS Alfa Aesar
44069AK
250mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA44069K2 SDS Alfa Aesar
44069K2
1L Each for $96.49
Specifications Description & Specifications

Chemical Identifiers

Liquid
MFCD00011137
Nickel plating solution

Specifications

Liquid
Ammonia-like
250mL
MFCD00011137
Green
Nickel plating solution

Safety and Handling

H334-H350-H360-H372-H317-H341-H302-H332-H411-H401: May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. May cause an allergic skin reaction. Suspected of causing genetic defects. Harmful if swallowed. Harmful if inhaled. Toxic to aquatic life with long lasting effects. Toxic to aquatic life.

DOTInformation : DOT Class: 9, Packaging Group: III