Please login to your online account to display your discounted pricing

Amberlite IRN-150, Ion Exchange Resin, Alfa Aesar

Styrene-DVB; Mix of strongly acidic and basic gel type resin

$46.26 - $565.76

Chemical Identifiers

Chemical Name or Material Amberlite IRN-150
Physical Form Beads
Molecular Formula Styrene-DVB
MDL Number MFCD00145822
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40992A9

  SDS
alfa aesar
40992A9
10kg Each for $565.76

AA4099236

  SDS
alfa aesar
4099236*
500g Each for $46.26
Description & Specifications

Specifications

Quantity 10kg
Color Amber
Density 1.1-1.3g/cm3
Physical Form Beads
MDL Number MFCD00145822
Odor Odorless
Molecular Formula Styrene-DVB
Appearance Amber beads
Ignition Point 500°C (932°F)
Chemical Name or Material Amberlite IRN-150