missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Amberlite™ IRA-67, ion exchange resin, Thermo Scientific Chemicals

Acrylic-DVB; Weakly basic gel type resin

$60.81 - $398.93

Chemical Identifiers

CAS 80747-90-6
MDL Number MFCD00145567
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4225330
View Documents Promotion Details
Thermo Scientific Chemicals
04225330
250 g
Each for $60.81
 
AA42253A1
View Documents Promotion Details
Thermo Scientific Chemicals
042253A1
1 kg
Each for $149.03
 
AA42253A7
View Documents Promotion Details
Thermo Scientific Chemicals
042253A7
5 kg
Each for $398.93
 
Description

Description

Amberlite™ IRA-67 is used for de-acidification and desalination of water and for isolation of acidic natural products. It allows for effective adsorption and desorption of naturally occurring organic molecules such as humic and fulvic acids that are present in many water supplies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

80747-90-6
MFCD00145567

Specifications

Beads
Amine-like
MFCD00145567
Tan
Insoluble in water.
80747-90-6
Acrylic-DVB
1.06g/cm3
250 g
Amberlite™ IRA-67, Ion Exchange Resin
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only