Thermo Scientific™ Quartz microscope slide, fused, 76.2x25.4x1.0mm (3.0x1.0x0.0394in)
Fused quartz slides manufactured by fusing naturally occurring crystalline silica
Supplier: Thermo Scientific™ 042297KG
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- High chemical purity, melting point and corrosion resistance
- Extremely hard with low coefficient of thermal expansion
- Features electrical insulation qualities and optical transmission from ultra-violet to infra-red
RUO â Research Use Only