Alexa Fluor™ 647-Poly-GT Peptide Substrate DFS Item
Alexa Fluor™ 647-Poly-GT Peptide Substrate
Manufacturer:  Life Technologies LSPV5836
 View more versions of this product
Catalog No. PV5836
Description
Description
A general AlexaFluor647-labeled peptide substrate consisting of a polymer of glutamate, and tyrosine that is used to monitor tyrosine kinase acitivty.
For use in combination with the LanthaScreen™ Eu-PY20 Antibody (PV5696) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
10nmol
–20°C
Far-Red
10nmol
Kinase
Alexa Fluor™ 647-Poly-GT Peptide Substrate, 10nmol, 30μM Concentration, General Alexa Fluor647-labeled Peptide Substrate Consisting of a Polymer of Glutamate, and Tyrosine that is used to Monitor Tyrosine Kinase Activity
Pharma and biopharma, target and lead identification and validation, combination with the LanthaScreen™ Eu-PY20 Antibody (PV5696) to evaluate inhibitors in medium to high throughput screening applications, drug discovery and development, kinase biology, biochemical kinase assays, LanthaScreen™ kinase activity assays
Alexa Fluor™ 647-Poly-GT Peptide Substrate
Documents
For Research Use Only. Not for use in diagnostic procedures.