Please login to your online account to display your discounted pricing

Aldehydes Mixture, 1000μg/mL in water, Ultra Scientific

$89.89 - $89.89

Chemical Identifiers

Chemical Name or Material Aldehyde mixture
CAS 007732-18-5
CAS Max % 99.8
CAS 000075-07-0
CAS Max % 0.1
CAS 000050-00-0
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-ALD-100-1

 
ultra scientific
801175000310
1 x 1mL Each for $89.89
Description & Specifications

Specifications

Quantity 1 x 1mL
Analyte or Component Names Acetaldehyde,formaldehyde,water
Solvent or Matrix Water
Concentration or Composition (by Analyte or Components) 1000μg/mL
CAS 007732-18-5
CAS Max % 0.1
Chemical Name or Material Aldehyde mixture
CAS Max % 0.1
CAS Max % 99.8
CAS 000075-07-0
CAS 000050-00-0