missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alcoholic Sulfuric Acid, for Phosphorus, Ricca Chemical

Chemical Identifiers

CAS 67-56-1,7664-93-9
CAS Min % 94.0,4.47
CAS Max % 97.0,4.65
Physical Form Liquid
Chemical Name or Material Alcoholic Sulfuric Acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
350-32 Ricca Chemical Company
RI35032
Poly natural 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R0350000500 Ricca Chemical Company
RIR0350000500A
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

For Phosphorus
Description & Specifications

Chemical Identifiers

67-56-1,7664-93-9
97.0,4.65
Alcoholic Sulfuric Acid
94.0,4.47
Liquid

Specifications

Liquid
1L
94.0,4.47
Poly natural
Clear colorless
67-56-1,7664-93-9
4.65,97.0
Alcoholic Sulfuric Acid