missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alcoholic Sulfuric Acid, For Phosphorus, LabChem™

$117.54 - $117.54

Chemical Identifiers

CAS 67-56-1,7664-93-9
CAS Max % 98,2
Linear Formula H2SO4
Formula Weight 98.08
UN Number UN2924
Chemical Name or Material Alcoholic Sulfuric Acid, For Phosphorus
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC105702 Lab Chem
LC105702
1L Each for $117.54
Specifications Description & Specifications

Chemical Identifiers

67-56-1,7664-93-9
H2SO4
UN2924
98,2
98.08
Alcoholic Sulfuric Acid, For Phosphorus

Specifications

98.08
Poly Bottle
Clear, colorless
7664-93-9,67-56-1
2,98
H2SO4
UN2924
Carbon monoxide Carbon dioxide May release flammable gases
Soluble in water
1L
Liquid
Room Temperature
Passes Test
Alcoholic Sulfuric Acid, For Phosphorus

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer (inhalation).
Causes damage to organs (central nervous system, kidney, liver, optic nerve).

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Use only outdoors or in a well-ventilated area.
Do not breathe mist, spray, vapors.
Do not eat, drink or smoke when using this product.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Immediately call a poison center or doctor/physician.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep at rest in a position comfortable for breathing.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use dry extinguishing powder, alcohol-resistant foam, carbon dioxide to extinguish.
Wash contaminated clothing before reuse.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

Danger