Please login to your online account to display your discounted pricing

Alcohol, Reagent, 50% (v/v) Aqueous Solution, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR254650020F

 View more versions of this product

Catalog No. R254650020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 20L
CAS Max % 46.0
CAS Min % 49.0
CAS Min % 2.0
Chemical Name or Material Alcohol, Reagent
Concentration or Composition Notes 50% (v/v)
Solvent or Matrix Aqueous Solution
CAS Max % 51.0
CAS Max % 3.0
CAS 67-56-1
CAS 64-17-5
CAS Min % 45.0
Packaging Cubitainer
CAS 67-63-0
CAS Min % 2.0
CAS Max % 3.0
CAS 7732-18-5

Specifications

Chemical Name or Material Alcohol, Reagent
CAS 7732-18-5
CAS Min % 49.0
CAS Max % 51.0
CAS 64-17-5
CAS Min % 45.0
CAS Max % 46.0
CAS 67-63-0
CAS Min % 2.0
CAS Max % 3.0
CAS 67-56-1
CAS Min % 2.0
CAS Max % 3.0
Physical Form Liquid