Please login to your online account to display your discounted pricing

Alcohol, Reagent, 20% (v/v), Fisher Chemical

Chemical Identifiers

Chemical Name or Material Reagent Alcohol, 20% (v/v)
CAS 7732-18-5
CAS Min % 79.0
CAS Max % 81.0
CAS 64-17-5
CAS Min % 18.0
CAS Max % 20.0
CAS 67-56-1
CAS Min % 0.8
CAS Max % 0.9
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SA9320-1

 
ricca chemical company
RISA93201
Poly Bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SA9320-4

  SDS
ricca chemical company
SA93204
Poly Bottle 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1L
DOT Information DOT Class 3, : Flammable Liquid
Methods For AOAC Method 992.22
Chemical Name or Material Reagent Alcohol, 20% (v/v)
CAS 7732-18-5
CAS Min % 79.0
CAS Max % 20.0
CAS 67-56-1
CAS Max % 0.9
CAS 67-63-0
CAS Min % 0.9
CAS Max % 1.0
Packaging Poly Bottle
CAS Max % 81.0
ChemAlert Storage Symbol Red
CAS Min % 18.0
CAS Min % 0.8
CAS 64-17-5
Safety and Handling