Please login to your online account to display your discounted pricing

Alcohol, OmniSolv™ Reagent (HPLC), EMD Millipore

Components: 90% (w/v) Ethyl Alcohol (CAS Reg. 64-17-5) + 5% (w/v) Methanol (CAS Reg. 67-56-1) + 5% (w/v) 2-Propanol (CAS Reg. 67-63-0)

$104.86 - $104.86

Chemical Identifiers

Chemical Name or Material Alcohol, OmniSolv™ Reagent
Name Note OmniPure™
Purity Grade HPLC Grade
Physical Form Liquid
Molecular Formula C2H5OH
Linear Formula CH3CH2OH
Formula Weight 46.7
Synonym Denatured Alcohol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MAX04456

 
emd millipore corporation
AX04456
Glass Bottle 1 liter Each for $104.86
Description & Specifications

Specifications

Purity Grade HPLC Grade
Formula Weight 46.7
Quantity 1 liter
Color Clear
DOT Information DOT Class 3, : Flammable Liquid
Concentration or Composition (by Analyte or Components) Assay
  • Ethanol 89.5-91.5% 2-Propanol 4.0-6.0% Methanol 3.5-5.5% Color (APHA)
  • ≤10 Residue after Evaporation
  • ≤4ppm Titrable Acid
  • ≤0.3μEq/g Titrable Base
  • ≤0.04μEq/g Optical Absorbance at 205nm
  • ≤1.00at 210nm
  • ≤0.65at 220nm
  • ≤0.35at 230nm
  • ≤0.20at 250nm
  • ≤0.04at 270nm
  • ≤0.01at 300nm
  • ≤0.005 Water
  • ≤0.04%Filtered through a 0.2 micron filter To pass test
Physical Form Liquid
Synonym Denatured Alcohol
Chemical Name or Material Alcohol, OmniSolv™ Reagent
Packaging Glass Bottle
Cleanup Symbol Solvent
Name Note OmniPure™
Molecular Formula C2H5OH
Linear Formula CH3CH2OH
Recommended Storage Room Temperature
Safety and Handling

Room Temperature