Please login to your online account to display your discounted pricing

Alcohol-Glycerin, 50-50 (Reagent Alcohol and Glycerin), LabChem™

$32.34 - $32.34

Chemical Identifiers

Chemical Name or Material Alcohol-Glycerin
CAS 56-81-5
CAS Max % 66.39
CAS 64-17-5
CAS Max % 30.25
CAS 67-63-0
CAS Max % 1.68
CAS 67-56-1
CAS Max % 1.68
Name Note 50-50 (Reagent Alcohol and Glycerin)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC105657

 
lab chem
LC105657
Poly Bottle 125mL Each for $32.34
Description & Specifications

Specifications

Formula Weight 92.09
Chemical Name or Material Alcohol-Glycerin
Identification Passes Test
CAS Max % 1.68
CAS 67-56-1
CAS Max % 1.68
Packaging Poly Bottle
Solubility Soluble in water
Storage Information Room Temperature
Name Note 50-50 (Reagent Alcohol and Glycerin)
Decomposition Information Carbon monoxide Carbon dioxide May release flammable gases
Physical Form Liquid
Color Clear, colorless
Quantity 125mL
Linear Formula HOCH2CH(OH)CH2OH
CAS 56-81-5
UN Number UN1987
Health Hazard 1 Danger
CAS Max % 66.39
Health Hazard 2 GHS H Statement
Flammable liquid and vapor.
Causes skin and serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).
CAS 64-17-5
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 30.25
CAS 67-63-0
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Use only outdoors or in a well-ventilated area.
Do not eat, drink or smoke when using this product.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Flammable liquid and vapor.
Causes skin and serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).