missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Albumin, Human, Fraction V Powder, 5g, MP Biomedicals

Used to eliminate background interference in ELISAs or other enzyme assay systems. Albumin, Human, Fraction V Powder, 5g, MP Biomedicals is used as standards in protein calibration studies.

Manufacturer:  Mp Biomedicals 08823234

Catalog No. ICN823234



Description

Description

  • Used in absorption, distribution, metabolism, and excretion (ADME) pharmacological research
  • Applications:
  • Cell culture
  • Drug delivery research
  • Cryopreservation of cells
  • Solubilize lipids in tissue culture
  • Blocking agents in Western blots or ELISA applications
Specifications

Specifications

Immunoassays, Mammalian Cell Culture
Human Albumin Fraction V
Modification of the Chen low pH Charcoal method
Plasma
Powder
7.2
5g
2° to 8°C
Documents