missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically Purified

For research use

$55.97 - $55.97

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements 2° to 8°C
Source Bovine Serum
Product Type Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6540009
View Documents
Alfa Aesar™
J6540009
10g Each for $55.97
 
Specifications

Specifications

Lyophilized powder
2° to 8°C
Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically purified
Suitable for immunoassays. Reduced fatty acid content minimizes non-specific binding of antibodies
No
Bovine Serum
Lyophilized Powder
RUO

For Research Use Only